The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017252.2 | NP_058948.1 | 1 - 1 | 281 | 124 |
GenBank mRNA | M84645.1 | - | 1 - 1 | 281 | 124 |
Z11834.1 | - | 1 - 1 | 253 | 124 |