Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vtn
Entrez Gene ID
29169
Gene Name
vitronectin
Gene Aliases
Aa1018, Vn
Chromosome Location
Chr.10: 65767960 - 65771040 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019156.2 NP_062029.2 6 - 7 1055 91
GenBank mRNA AY318963.1 - - 1052 91
BC105821.1 - 6 - 7 1055 91
FQ209609.1 - 6 - 7 1068 91
FQ209682.1 - 6 - 7 1063 91
FQ210146.1 - 4 - 5 675 91
FQ211325.1 - 2 - 3 189 91
FQ212575.1 - 6 - 7 1055 91
FQ218318.1 - 6 - 7 1067 91
FQ218401.1 - 4 - 5 628 91
FQ219297.1 - 6 - 7 1070 91
U44845.1 - 6 - 7 1045 91

View Full Product Details