The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020542.2 | NP_065417.1 | 2 - 2 | 1006 | 96 |
GenBank mRNA | BC079207.1 | - | 2 - 2 | 1006 | 96 |