The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021596.2 | NP_067607.1 | 1 - 2 | 123 | 69 |
GenBank mRNA | AF128531.1 | - | 1 - 2 | 125 | 69 |