The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021702.1 | NP_067734.1 | 7 - 8 | 719 | 65 |
XM_006240485.2 | XP_006240547.1 | 8 - 9 | 755 | 65 | |
XM_006240486.2 | XP_006240548.1 | 8 - 9 | 752 | 65 | |
XM_006240487.2 | XP_006240549.1 | 8 - 9 | 755 | 65 | |
XM_006240488.2 | XP_006240550.1 | 7 - 8 | 671 | 65 | |
XM_006240490.1 | XP_006240552.1 | 8 - 9 | 781 | 65 | |
XM_006240493.2 | XP_006240555.1 | 7 - 8 | 674 | 65 | |
GenBank mRNA | Y12319.1 | - | 7 - 8 | 719 | 65 |