The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021745.1 | NP_068513.1 | 3 - 4 | 609 | 95 |
XM_008765260.1 | XP_008763482.1 | 2 - 3 | 967 | 95 | |
XM_008765261.1 | XP_008763483.1 | 2 - 3 | 966 | 95 | |
XM_008765262.1 | XP_008763484.1 | 3 - 4 | 648 | 95 | |
GenBank mRNA | BC083815.1 | - | 2 - 3 | 487 | 95 |
U18374.1 | - | 3 - 4 | 609 | 95 |