The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022232.1 | NP_071568.1 | 3 - 4 | 324 | 81 |
GenBank mRNA | AB017702.3 | - | 3 - 4 | 324 | 81 |
BC061764.1 | - | 3 - 4 | 419 | 81 |