The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022253.1 | NP_071589.1 | 3 - 4 | 354 | 68 |
GenBank mRNA | AB038038.1 | - | 3 - 4 | 354 | 68 |