The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008759812.1 | XP_008758034.1 | 7 - 8 | 1778 | 89 |
GenBank mRNA | AF136401.1 | - | - | 1785 | 89 |