Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snap23
Entrez Gene ID
64630
Gene Name
synaptosomal-associated protein 23
Gene Aliases
-
Chromosome Location
Chr.3: 112333972 - 112362960 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022689.2 NP_073180.1 3 - 4 250 83
GenBank mRNA AF052596.1 - - 102 83
BC127494.1 - 3 - 4 250 83

View Full Product Details