The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022700.2 | NP_073191.1 | 4 - 5 | 508 | 60 |
XM_006231559.2 | XP_006231621.1 | 5 - 6 | 1029 | 60 | |
GenBank mRNA | BC084722.1 | - | 4 - 5 | 508 | 60 |
U12568.1 | - | 4 - 5 | 512 | 60 | |
X76921.1 | - | 5 - 6 | 640 | 60 |