Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cplx1
Entrez Gene ID
64832
Gene Name
complexin 1
Gene Aliases
-
Chromosome Location
Chr.14: 2194891 - 2226610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006250571.2 XP_006250633.1 1 - 2 126 94
GenBank mRNA D70817.1 - 1 - 2 35 94
U35098.1 - 1 - 2 33 94

View Full Product Details