Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpv4
Entrez Gene ID
66026
Gene Name
transient receptor potential cation channel, subfamily V, member 4
Gene Aliases
Otrpc4, Vroac
Chromosome Location
Chr.12: 47698915 - 47737902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023970.1 NP_076460.1 5 - 6 950 59
XM_006249466.2 XP_006249528.1 5 - 6 979 59
GenBank mRNA AF263521.1 - 5 - 6 950 59

View Full Product Details