The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030839.3 | NP_110466.2 | 3 - 4 | 550 | 71 |
XM_008762036.1 | XP_008760258.1 | 3 - 4 | 1144 | 71 | |
GenBank mRNA | AF313453.1 | - | 3 - 4 | 735 | 71 |
AF375466.1 | - | 3 - 4 | 550 | 71 |