The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030861.1 | NP_110488.1 | 1 - 2 | 36 | 85 |
XM_006246182.2 | XP_006246244.1 | 2 - 3 | 514 | 85 | |
XM_008767715.1 | XP_008765937.1 | 2 - 3 | 528 | 85 | |
GenBank mRNA | AB012874.1 | - | 2 - 3 | 503 | 85 |
AB012875.1 | - | 1 - 2 | 54 | 85 | |
D16302.1 | - | 1 - 2 | 36 | 85 |