Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Gtf2b
Entrez Gene ID
81673
Gene Name
general transcription factor IIB
Gene Aliases
-
Chromosome Location
Chr.2: 248709433 - 248727741 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031041.1 NP_112303.1 3 - 3 142 76
GenBank mRNA BC085345.1 - 3 - 3 147 76
FQ229769.1 - 3 - 3 130 76
FQ231337.1 - 3 - 3 172 76
X65948.1 - 3 - 3 142 76

View Full Product Details