The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031699.2 | NP_113887.2 | 2 - 3 | 566 | 113 |
GenBank mRNA | AF195500.1 | - | 2 - 3 | 113 | |
BC061992.1 | - | 2 - 3 | 566 | 113 | |
FQ220868.1 | - | 2 - 3 | 535 | 113 |