The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033236.1 | NP_150239.1 | 9 - 10 | 860 | 153 |
GenBank mRNA | BC061542.1 | - | 9 - 10 | 902 | 153 |
D50694.1 | - | 9 - 10 | 860 | 153 | |
FQ213265.1 | - | 6 - 7 | 663 | 153 | |
FQ215767.1 | - | 6 - 7 | 663 | 153 | |
FQ225383.1 | - | 6 - 7 | 663 | 153 |