Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcnj13
- Entrez Gene ID
-
94341
- Gene Name
- potassium channel, inwardly rectifying subfamily J, member 13
- Gene Aliases
-
Kir7.1
- Chromosome Location
-
Chr.9: 94486719 - 94495333 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053608.2 |
NP_446060.1 |
1 - 2 |
97 |
113 |
View Full Product Details