The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053678.1 | NP_446130.1 | 2 - 3 | 549 | 83 |
GenBank mRNA | AF135839.1 | - | 2 - 3 | 549 | 83 |