Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyth3
Entrez Gene ID
116693
Gene Name
cytohesin 3
Gene Aliases
Pscd3, Sec7, Sec7C
Chromosome Location
Chr.12: 12877559 - 12907771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053912.2 NP_446364.2 - 161 81
GenBank mRNA BC101884.1 - - 161 81
U83897.1 - - 154 81

View Full Product Details