The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053963.2 | NP_446415.2 | 4 - 5 | 660 | 65 |
GenBank mRNA | BC088120.1 | - | 4 - 5 | 660 | 65 |
X98517.1 | - | 4 - 5 | 660 | 65 |