The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080696.2 | NP_542427.1 | 3 - 4 | 504 | 61 |
GenBank mRNA | AF361346.1 | - | 3 - 4 | 504 | 61 |