Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lta
Entrez Gene ID
25008
Gene Name
lymphotoxin alpha
Gene Aliases
Tnfb
Chromosome Location
Chr.20: 4852671 - 4854677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080769.2 NP_542947.1 3 - 4 344 86

View Full Product Details