The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080892.1 | NP_543168.1 | 6 - 7 | 909 | 65 |
GenBank mRNA | AB036799.1 | - | 6 - 7 | 909 | 65 |
BC074008.1 | - | 6 - 7 | 919 | 65 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008761279.1 | XP_008759501.1 | - | 909 | 65 |