Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmd9
Entrez Gene ID
161475
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 9
Gene Aliases
Bridge, Bridge-1
Chromosome Location
Chr.12: 38786539 - 38805121 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130430.1 NP_569114.1 5 - 6 1134 66
GenBank mRNA AF067728.1 - 5 - 6 1134 66
FQ214178.1 - - 678 66
FQ225583.1 - - 667 66
FQ227319.1 - - 667 66
FQ229304.1 - - 700 66

View Full Product Details