The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130821.1 | NP_570834.1 | 2 - 3 | 133 | 102 |
GenBank mRNA | AB067679.1 | - | 2 - 3 | 133 | 102 |