The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133526.1 | NP_598210.1 | 5 - 6 | 678 | 65 |
GenBank mRNA | BC061785.1 | - | 5 - 6 | 534 | 65 |
FQ214867.1 | - | 5 - 6 | 586 | 65 | |
FQ215876.1 | - | 1 - 2 | 50 | 65 | |
FQ216712.1 | - | 5 - 6 | 707 | 65 | |
FQ220073.1 | - | 1 - 2 | 50 | 65 | |
FQ220093.1 | - | 1 - 2 | 50 | 65 | |
FQ227464.1 | - | 5 - 6 | 695 | 65 | |
FQ229225.1 | - | 1 - 2 | 50 | 65 | |
FQ229522.1 | - | 5 - 6 | 596 | 65 | |
Y13275.2 | - | 5 - 6 | 678 | 65 |