Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cx3cr1
Entrez Gene ID
171056
Gene Name
chemokine (C-X3-C motif) receptor 1
Gene Aliases
Rbs11
Chromosome Location
Chr.8: 128738689 - 128755376 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133534.1 NP_598218.1 1 - 2 61 76
GenBank mRNA U04808.1 - 1 - 2 61 76

View Full Product Details