The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138841.1 | NP_620196.1 | 1 - 2 | 687 | 72 |
GenBank mRNA | AF208288.1 | - | 1 - 2 | 687 | 72 |