Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asrgl1
Entrez Gene ID
246307
Gene Name
asparaginase like 1
Gene Aliases
Gliap, Hiob2
Chromosome Location
Chr.1: 225309737 - 225329743 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145089.4 NP_659557.1 1 - 2 108 62
GenBank mRNA AF329099.1 - 1 - 2 108 62
AJ427914.2 - - 122 62
FQ212762.1 - 1 - 2 82 62

View Full Product Details