The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145880.3 | NP_665887.3 | 1 - 2 | 770 | 64 |
GenBank mRNA | S71523.1 | - | 1 - 2 | 770 | 64 |