The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172336.3 | NP_758839.3 | 1 - 2 | 197 | 68 |
XM_006228980.1 | XP_006229042.1 | 2 - 3 | 475 | 68 | |
GenBank mRNA | AY123225.1 | - | 1 - 2 | 197 | 68 |
BC061786.1 | - | 2 - 3 | 475 | 68 |