Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chst15
Entrez Gene ID
286974
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases
GalNAc4S6ST, Galnac4s-6st
Chromosome Location
Chr.1: 204243396 - 204328970 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173310.3 NP_775432.3 3 - 4 1049 72
XM_006230430.2 XP_006230492.1 4 - 5 2616 72
XM_006230431.2 XP_006230493.1 4 - 5 3356 72
XM_006230433.2 XP_006230495.1 4 - 5 2851 72
XM_008759868.1 XP_008758090.1 4 - 5 1612 72
XM_008759869.1 XP_008758091.1 4 - 5 3355 72
GenBank mRNA AB086953.1 - 3 - 4 1049 72
BC085687.1 - 3 - 4 1310 72

View Full Product Details