Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ceacam10
- Entrez Gene ID
-
287009
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 81643809 - 81650085 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173339.1 |
NP_775461.1 |
2 - 3 |
503 |
127 |
GenBank mRNA |
U23056.1 |
- |
2 - 3 |
503 |
127 |
- Gene Symbol
- LOC103689942
- Entrez Gene ID
-
103689942
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 83003835 - 83010148 on Build Rnor_6.0
View Full Product Details