The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175837.1 | NP_787031.1 | 2 - 3 | 374 | 69 |
XM_006238712.2 | XP_006238774.1 | 2 - 3 | 386 | 69 | |
GenBank mRNA | BC089761.1 | - | 2 - 3 | 352 | 69 |
X07259.1 | - | 2 - 3 | 344 | 69 |