Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grsf1
Entrez Gene ID
305256
Gene Name
G-rich RNA sequence binding factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 21039990 - 21056414 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100890.1 NP_001094360.1 4 - 5 827 136
XM_006250775.2 XP_006250837.1 4 - 5 714 136
GenBank mRNA CK601425.1 - 2 - 3 243 136

View Full Product Details