The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012818.2 | NP_036950.1 | 3 - 4 | 423 | 63 |
XM_006247792.1 | XP_006247854.1 | 3 - 4 | 1419 | 63 | |
XM_006247793.1 | XP_006247855.1 | 3 - 4 | 1414 | 63 | |
GenBank mRNA | DQ075321.1 | - | 3 - 4 | 611 | 63 |
U29664.1 | - | 2 - 3 | 212 | 63 | |
U38306.1 | - | 3 - 4 | 423 | 63 | |
U40803.1 | - | 3 - 4 | 510 | 63 |