Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkaa1
Entrez Gene ID
65248
Gene Name
protein kinase, AMP-activated, alpha 1 catalytic subunit
Gene Aliases
AMPKalpha1
Chromosome Location
Chr.2: 54857688 - 54893404 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019142.2 NP_062015.2 2 - 3 269 92
GenBank mRNA FN804412.1 - 2 - 3 247 92
U40819.1 - 2 - 3 236 92

View Full Product Details