The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031325.1 | NP_112615.1 | 10 - 11 | 1365 | 100 |
GenBank mRNA | AB013732.1 | - | 10 - 11 | 1365 | 100 |