Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ighmbp2
Entrez Gene ID
29532
Gene Name
immunoglobulin mu binding protein 2
Gene Aliases
AEP
Chromosome Location
Chr.1: 218509274 - 218531922 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031586.1 NP_113774.1 2 - 3 257 104
GenBank mRNA AF199411.1 - 2 - 3 257 104
BC099790.1 - 2 - 3 453 104

View Full Product Details