The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031616.1 | NP_113804.1 | 8 - 9 | 119 | |
GenBank mRNA | AF013965.1 | - | 8 - 9 | 119 | |
AF013967.1 | - | 8 - 9 | 119 | ||
BC087012.1 | - | 8 - 9 | 779 | 120 | |
FQ211774.1 | - | 8 - 9 | 819 | 120 | |
FQ211917.1 | - | 8 - 9 | 771 | 120 |