Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctcf
Entrez Gene ID
83726
Gene Name
CCCTC-binding factor (zinc finger protein)
Gene Aliases
-
Chromosome Location
Chr.19: 37600377 - 37649674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031824.1 NP_114012.1 2 - 3 158 69
XM_006255536.1 XP_006255598.1 1 - 2 108 69
GenBank mRNA AF133731.2 - 2 - 3 194 69
FQ227933.1 - 2 - 3 352 69
FQ228237.1 - 2 - 3 286 69

View Full Product Details