Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfm1
Entrez Gene ID
93667
Gene Name
olfactomedin 1
Gene Aliases
D2Sut1e, Noe1
Chromosome Location
Chr.3: 6761136 - 6798740 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053573.1 NP_446025.1 1 - 2 439 119
XM_006233876.2 XP_006233938.1 1 - 2 467 119
GenBank mRNA U03415.1 - 1 - 2 439 119
U03417.1 - 1 - 2 439 119

View Full Product Details