Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnase1l3
Entrez Gene ID
116687
Gene Name
deoxyribonuclease 1-like 3
Gene Aliases
-
Chromosome Location
Chr.15: 18710492 - 18736476 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053907.1 NP_446359.1 6 - 7 918 85
GenBank mRNA AF039852.1 - 6 - 7 868 85
BC088122.1 - 6 - 7 916 85
FQ226037.1 - 6 - 7 745 85
FQ227612.1 - 6 - 7 745 85
FQ230283.1 - 6 - 7 745 85
FQ230990.1 - 6 - 7 745 85
FQ231646.1 - 6 - 7 744 85
FQ231870.1 - 6 - 7 748 85
FQ231939.1 - 6 - 7 745 85
FQ231976.1 - 6 - 7 801 85
FQ233228.1 - 6 - 7 801 85
U75689.1 - 6 - 7 918 85

Gene Symbol
LOC103690049
Entrez Gene ID
103690049
Gene Name
deoxyribonuclease gamma-like
Gene Aliases
-
Chromosome Location
Chr.15: 17927877 - 17945116 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770646.1 XP_008768868.1 4 - 5 653 85

View Full Product Details