Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcna
Entrez Gene ID
25737
Gene Name
proliferating cell nuclear antigen
Gene Aliases
PCNAR, Pcna/cyclin
Chromosome Location
Chr.3: 124880698 - 124884570 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022381.3 NP_071776.1 4 - 5 644 116
GenBank mRNA BC060570.1 - 4 - 5 729 116
FQ219845.1 - 4 - 5 725 116
FQ219897.1 - 4 - 5 725 116
FQ220101.1 - 4 - 5 725 116
FQ220835.1 - 4 - 5 725 116
FQ220924.1 - 4 - 5 724 116
FQ225620.1 - 3 - 4 349 116
FQ226726.1 - 4 - 5 726 116
FQ227521.1 - 4 - 5 725 116
FQ229469.1 - 4 - 5 725 116
FQ229972.1 - 4 - 5 726 117
FQ230190.1 - 4 - 5 725 116
FQ232129.1 - 4 - 5 738 116
FQ233125.1 - 4 - 5 725 116
FQ233579.1 - 4 - 5 725 116
FQ233966.1 - 4 - 5 725 116
FQ234183.1 - 4 - 5 727 116
FQ234500.1 - 4 - 5 725 116
FQ234529.1 - 4 - 5 725 116
FQ234547.1 - 4 - 5 725 116
FQ234986.1 - 4 - 5 725 116
FQ235296.1 - 4 - 5 725 116
Y00047.1 - 4 - 5 644 116

View Full Product Details