Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkcz
Entrez Gene ID
25522
Gene Name
protein kinase C, zeta
Gene Aliases
14-3-3-zetaisoform, Pkcz, r14-3-3
Chromosome Location
Chr.5: 172658071 - 172769492 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022507.1 NP_071952.1 17 - 18 1840 64
XM_006239535.2 XP_006239597.1 14 - 15 4335 64
XM_006239536.2 XP_006239598.1 14 - 15 3541 64
XM_008764337.1 XP_008762559.1 17 - 18 1741 64
GenBank mRNA J04532.1 - 17 - 18 1840 64
M18332.1 - 14 - 15 1393 64
U27191.1 - 14 - 15 1605 64

View Full Product Details