Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sult1b1
Entrez Gene ID
64305
Gene Name
sulfotransferase family, cytosolic, 1B, member 1
Gene Aliases
ST1B1
Chromosome Location
Chr.14: 22142412 - 22155246 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022513.2 NP_071958.1 1 - 2 237 93
GenBank mRNA D89375.1 - 1 - 2 188 93
FQ209476.1 - 1 - 2 206 93
FQ209486.1 - 1 - 2 271 93
FQ209579.1 - 1 - 2 292 93
FQ212608.1 - 1 - 2 248 93
FQ218295.1 - 1 - 2 250 93
U38419.1 - 1 - 2 240 93

View Full Product Details