Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sfrp4
Entrez Gene ID
89803
Gene Name
secreted frizzled-related protein 4
Gene Aliases
-
Chromosome Location
Chr.17: 47383983 - 47394065 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053544.1 NP_445996.1 2 - 3 616 85
GenBank mRNA AF012891.1 - 2 - 3 616 85
AF140346.1 - 2 - 3 767 85
AF220608.1 - - 864 85
FQ229224.1 - 1 - 2 47 85

View Full Product Details