Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wif1
Entrez Gene ID
114557
Gene Name
Wnt inhibitory factor 1
Gene Aliases
-
Chromosome Location
Chr.7: 63094932 - 63165158 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053738.1 NP_446190.1 9 - 10 1297 113
XM_006241395.2 XP_006241457.1 9 - 10 1354 113
GenBank mRNA AY030278.1 - 9 - 10 1297 113
BC072473.1 - 9 - 10 1165 113

View Full Product Details